Learn More
MSC Envu Insecticide Dust, 1% Cyfluthrin, Ready-to-Use, Low Odor
Catalog No. 502835661
Encompass
This insecticide dust is a ready-to-use formulation containing 1% Cyfluthrin, designed for professional pest management. It offers effective residual control of various pests in difficult, hard-to-reach sites, featuring a low-odor and non-staining application.
  • Effective pest control with a long residual
  • Low odor and non-staining
  • Broad label directions for versatile application
  • Ready-to-use dust formulation
  • Contains cyfluthrin as the active ingredient
  • Labeled for indoor and outdoor use
  • For use by pest management professionals and commercial applicators
Catalog No. 50-283-5661 Supplier MSC Supplier No. 74335670
Add to Cart
Add to Cart
This insecticide dust is a ready-to-use formulation containing 1% Cyfluthrin, designed for professional pest management. It offers effective residual control of various pests in difficult, hard-to-reach sites, featuring a low-odor and non-staining application.
  • Effective pest control with a long residual
  • Low odor and non-staining
  • Broad label directions for versatile application
  • Ready-to-use dust formulation
  • Contains cyfluthrin as the active ingredient
  • Labeled for indoor and outdoor use
  • For use by pest management professionals and commercial applicators