Learn More
MSC BASF CORP Pressurized Lambda Insecticide, Residual, 14 oz Can, Lambda-cyhalothrin
Catalog No. 502837964
Encompass
This residual insecticide is an easy-to-use aerosol featuring lambda cyhalothrin. It is effective against a wide range of insects and is compatible with System III.
  • Features lambda cyhalothrin as the active ingredient.
  • Effective against a wide range of insects.
  • Easy-to-use aerosol for convenient application.
  • Compatible with System III.
  • Controls pests such as ants, bed bugs, cockroaches, spiders, and many more.
  • Suitable for various applications including homes, hospitals, and food processing plants.
Catalog No. 50-283-7964 Supplier MSC Supplier No. 74333352
This residual insecticide is an easy-to-use aerosol featuring lambda cyhalothrin. It is effective against a wide range of insects and is compatible with System III.
  • Features lambda cyhalothrin as the active ingredient.
  • Effective against a wide range of insects.
  • Easy-to-use aerosol for convenient application.
  • Compatible with System III.
  • Controls pests such as ants, bed bugs, cockroaches, spiders, and many more.
  • Suitable for various applications including homes, hospitals, and food processing plants.