Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Insects Rubbing Plates
SDPSCI_ED

Catalog No. S13216
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13216 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13216 Supplier Roylco™ Supplier No. R5803
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use these magical molded plates to create rubs and study insects.

  • Place a sheet of paper over the rubbing plate and rub with the side of a crayon to reveal the pattern underneath
  • Color in the details with markers or colored pencils
  • Fill with or press into modeling clay
  • Great with papier mâché!
  • Make sun catchers using white glue and food color
  • Simply fill a rubbing plate and let dry
  • Peel the resulting mold off, and you are done

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Age ≥3 Years
Height (English) 6.5 in.
Includes 16 Plates, Teaching Guide
Height (Metric) 17 cm
Length (Metric) 11 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.