Learn More
Microsolv Technology Corporation Insert 100Ul W/Springs
Catalog No. 5001452056
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 250ul fill volume with a conical point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52056 Supplier Microsolv Technology Corporation Supplier No. 9502S02E
Inserts, Low Volume for Vials, Glass. Clear, 250ul fill volume with a conical point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. MicroSolv Brand. 100/PK.