Learn More
Waters Corp Waters Low Volume Insert, 150 μL, Plastic Spring, 100/pk
Catalog No. 50819086 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This low volume insert provides an efficient solution for applications with limited sample volumes, enabling compatibility with various laboratory equipment. Designed to fit into standard vials and collection plates, users can select the appropriate insert based on their specific sample volume and vial size requirements.
  • 150 μL volume capacity.
  • Designed to fit into standard vials and collection plates for compatibility with various laboratory equipment.
  • Includes a plastic spring to secure the sample and prevent leakage.
  • Made of high-quality materials to ensure chemical compatibility and minimize sample contamination.
  • Each pack contains 100 inserts.
Catalog No. 50-819-086 Supplier Waters Corp Supplier No. WAT094171
This low volume insert provides an efficient solution for applications with limited sample volumes, enabling compatibility with various laboratory equipment. Designed to fit into standard vials and collection plates, users can select the appropriate insert based on their specific sample volume and vial size requirements.
  • 150 μL volume capacity.
  • Designed to fit into standard vials and collection plates for compatibility with various laboratory equipment.
  • Includes a plastic spring to secure the sample and prevent leakage.
  • Made of high-quality materials to ensure chemical compatibility and minimize sample contamination.
  • Each pack contains 100 inserts.