Learn More
Microsolv Technology Corporation Insert 250Ul Self Aligning
Catalog No. 5001452065
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a conical point and 6x31mm outer dimensions. Self aligning, for use in 2ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52065 Supplier Microsolv Technology Corporation Supplier No. 9502S03N
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a conical point and 6x31mm outer dimensions. Self aligning, for use in 2ml autosampler vials. AQ Brand. 100/PK.