Learn More
Microsolv Technology Corporation Insert 250Ul Silan Self Align
Catalog No. 5001452066
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 300ul fill volume with conical point and 6x31mm outer dimensions. Self aligning, for use in 2ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52066 Supplier Microsolv Technology Corporation Supplier No. 9502S03ND
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 300ul fill volume with conical point and 6x31mm outer dimensions. Self aligning, for use in 2ml autosampler vials. AQ Brand. 100/PK.