Learn More
Microsolv Technology Corporation Insert 300Ul Conical Point
Catalog No. 5001452055
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a wider, conical point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. AQ Brand. 1000/CS.
Catalog No. 50-014-52055 Supplier Microsolv Technology Corporation Supplier No. 9502S02CPK
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a wider, conical point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. AQ Brand. 1000/CS.