Learn More
Microsolv Technology Corporation Insert 300Ul Conical Pt Eco
Catalog No. 5001452054
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a wider, conical point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52054 Supplier Microsolv Technology Corporation Supplier No. 9502S02CP
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a wider, conical point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. AQ Brand. 100/PK.