Learn More
Waters Corp Waters Corporation Insert, 6 x 29 mm, 300 μL, Plastic Spring
Catalog No. 50818793 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Vial inserts provide a solution for applications with limited sample volume. These inserts are designed to help distribute solutions, making them accessible for needles and other applications. They include a plastic spring to facilitate sample extraction and analysis and can help prevent exposure of light or oxygen-sensitive samples.
  • Designed for limited sample volume applications.
  • Includes a plastic spring for easier sample extraction and analysis.
  • Helps prevent exposure for light or oxygen-sensitive samples.
  • Compatible with various vials, such as 12 x 32 mm screw neck vials.
  • Made from polypropylene, offering reduced weight, breakage, disposal costs, and non-reactivity with most sample chemistries.
Catalog No. 50-818-793 Supplier Waters Corp Supplier No. WAT094170
Vial inserts provide a solution for applications with limited sample volume. These inserts are designed to help distribute solutions, making them accessible for needles and other applications. They include a plastic spring to facilitate sample extraction and analysis and can help prevent exposure of light or oxygen-sensitive samples.
  • Designed for limited sample volume applications.
  • Includes a plastic spring for easier sample extraction and analysis.
  • Helps prevent exposure for light or oxygen-sensitive samples.
  • Compatible with various vials, such as 12 x 32 mm screw neck vials.
  • Made from polypropylene, offering reduced weight, breakage, disposal costs, and non-reactivity with most sample chemistries.