Learn More
Waters Corp Waters Corporation Low Volume Glass Insert, 150 μL, 100 per Pack
Catalog No. 50819162 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Vial inserts provide a solution for applications with limited sample volume, allowing for the extraction of essential samples. This low volume glass insert is designed for HPLC instrument platforms and suitable for various applications, including biopharmaceutical, environmental, energy and fuels, forensics, materials science, proteomics, and metabolomics sectors. Proper selection ensures longer preservation of valuable samples by extracting only the necessary amount.
  • Provides a solution for applications with limited sample volume.
  • Enables extraction of the most essential samples.
  • Designed and suited for HPLC instrument platforms and a wide range of applications.
  • Ensures longer preservation of valuable samples by extracting only the necessary amount.
  • Suitable for use in biopharmaceutical, environmental, energy and fuels, forensics, materials science, proteomics, and metabolomics sectors.
Catalog No. 50-819-162 Supplier Waters Corp Supplier No. WAT072294
Vial inserts provide a solution for applications with limited sample volume, allowing for the extraction of essential samples. This low volume glass insert is designed for HPLC instrument platforms and suitable for various applications, including biopharmaceutical, environmental, energy and fuels, forensics, materials science, proteomics, and metabolomics sectors. Proper selection ensures longer preservation of valuable samples by extracting only the necessary amount.
  • Provides a solution for applications with limited sample volume.
  • Enables extraction of the most essential samples.
  • Designed and suited for HPLC instrument platforms and a wide range of applications.
  • Ensures longer preservation of valuable samples by extracting only the necessary amount.
  • Suitable for use in biopharmaceutical, environmental, energy and fuels, forensics, materials science, proteomics, and metabolomics sectors.