Learn More
Sarstedt Inc Tube Inserts for CryoPure Tubes, Red, 100/Pack
Catalog No. 50873810 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Insert; CyroPure; Sarstedt; Color: red; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; 100/pk.
Catalog No. 50-873-810 Supplier Sarstedt Inc Supplier No. 65.386.002
Tube, Insert; CyroPure; Sarstedt; Color: red; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; 100/pk.