Learn More
Bellco Glass, Inc. Insert Flat Bottom .25mL 5x31mm
SDP
Catalog No. 501945407
Encompass_Preferred
Inserts are pre-assembled with a polymer bottom spring. The polymer spring acts as a shock absorbent that protects against breakage if the needle bottoms out. The conical design permits complete sample removal. MADE IN THE USA. Flat bottom Size: .25ml
Catalog No. 50-194-5407 Supplier Bellco Glass, Inc. Supplier No. 207004300
Inserts are pre-assembled with a polymer bottom spring. The polymer spring acts as a shock absorbent that protects against breakage if the needle bottoms out. The conical design permits complete sample removal. MADE IN THE USA. Flat bottom Size: .25ml