Learn More
Chemglass Life Sciences Insert, 0.15mL, Glass, LV with Bottom Spring
SDP
Catalog No. 501532493
Encompass_Preferred
Insert, 0.15mL, Glass, LV with Bottom Spring. Glass inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog No. 50-153-2493 Supplier Chemglass Life Sciences Supplier No. CV14110529
Insert, 0.15mL, Glass, LV with Bottom Spring. Glass inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.