Learn More
Chemglass Life Sciences Insert, 0.1mL, Glass, LV with Plastic Flange
SDP
Catalog No. 501532491
Encompass_Preferred
Insert, 0.1mL, Glass, LV with Plastic Flange. Glass inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog No. 50-153-2491 Supplier Chemglass Life Sciences Supplier No. CV14050529
Insert, 0.1mL, Glass, LV with Plastic Flange. Glass inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.