Learn More
Chemglass Life Sciences Insert, 0.25mL, Glass, BM with Glass Flange
SDP
Catalog No. 501532495
Encompass_Preferred
Insert, 0.25mL, Glass, BM with Glass Flange. Both glass and plastic inserts have been engineered to fit the large opening in vials containing a nominal I.D. of 6mm. Inserts are available assembled with a polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog No. 50-153-2495 Supplier Chemglass Life Sciences Supplier No. CV14210631
Insert, 0.25mL, Glass, BM with Glass Flange. Both glass and plastic inserts have been engineered to fit the large opening in vials containing a nominal I.D. of 6mm. Inserts are available assembled with a polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.