Learn More
Chemglass Life Sciences Glass Insert, 0.25mL, BM, No Spring
SDP
Catalog No. 501532494
Encompass_Preferred
Glass Insert, 0.25mL, BM, No Spring. Due to the conical design of the snap plug, limited volume inserts used in the shell vial stay centered and straight allowing for easy sample removal.
Catalog No. 50-153-2494 Supplier Chemglass Life Sciences Supplier No. CV14180629
Glass Insert, 0.25mL, BM, No Spring. Due to the conical design of the snap plug, limited volume inserts used in the shell vial stay centered and straight allowing for easy sample removal.