Learn More
Chemglass Life Sciences Insert Glass .05mL BM w/Bottom Spring, 6x25mm, Silanized
SDP
Supplier:  Chemglass Life Sciences CV1402S625
Encompass_Preferred
Insert Glass .05mL BM w/Bottom Spring, 6x25mm, Silanized. Both glass and plastic inserts have been engineered to fit the large opening in vials containing a nominal I.D. of 6mm. Inserts are available assembled with a polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog No. 50-153-2490