Learn More
Microsolv Technology Corporation Insert Ms Plastic Spng Pk
Catalog No. 5001452062
Encompass_Preferred
Inserts, Low Volume for Vials, Plastic. Translucent, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. LCMS Compatible. AQ Brand. 100/PK.
Catalog No. 50-014-52062 Supplier Microsolv Technology Corporation Supplier No. 9502S02PP
Inserts, Low Volume for Vials, Plastic. Translucent, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. LCMS Compatible. AQ Brand. 100/PK.