Learn More
Chemglass Life Sciences Insert, 0.1mL, Polypropylene, LV, No Spring
SDP
Catalog No. 501532492
Encompass_Preferred
Insert, 0.1mL, Polypropylene, LV, No Spring. Plastic inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog No. 50-153-2492 Supplier Chemglass Life Sciences Supplier No. CV14060530
Insert, 0.1mL, Polypropylene, LV, No Spring. Plastic inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.