Learn More
Chemglass Life Sciences Insert, 0.1mL, Polypropylene, LV with Bottom Spring
SDP
Catalog No. 50230831
Encompass_Preferred
Insert, 0.1mL, Polypropylene, LV with Bottom Spring. Both glass and plastic inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog No. 50-230-831 Supplier Chemglass Life Sciences Supplier No. CV14070530
Insert, 0.1mL, Polypropylene, LV with Bottom Spring. Both glass and plastic inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.