Learn More
Waters Corp Waters Corporation Insert, Polypropylene, 200 μL, 8 x 40 mm Vial
Catalog No. 50780712 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polypropylene insert with polyspring is designed for applications with limited sample volume, enabling maximum sample extraction. Suitable for HPLC instrument platforms across pharmaceutical, environmental, energy and fuels, forensics, materials science, biopharmaceutical, proteomics, and metabolomics industries. Its plastic springs offer shock-absorbing function to prevent syringe damage. Manufactured from inert polypropylene, it prevents compound adhesion common with polar glass surfaces, ensuring pure analysis.
  • Maximum extraction of limited samples.
  • Compatible with HPLC platforms and diverse applications (pharmaceutical, environmental, energy & fuels, forensics, materials science, biopharmaceutical, proteomics, metabolomics).
  • Plastic springs provide shock absorption, preventing syringe damage.
  • Polypropylene construction prevents compound adhesion to surfaces.
Catalog No. 50-780-712 Supplier Waters Corp Supplier No. 186001728
This polypropylene insert with polyspring is designed for applications with limited sample volume, enabling maximum sample extraction. Suitable for HPLC instrument platforms across pharmaceutical, environmental, energy and fuels, forensics, materials science, biopharmaceutical, proteomics, and metabolomics industries. Its plastic springs offer shock-absorbing function to prevent syringe damage. Manufactured from inert polypropylene, it prevents compound adhesion common with polar glass surfaces, ensuring pure analysis.
  • Maximum extraction of limited samples.
  • Compatible with HPLC platforms and diverse applications (pharmaceutical, environmental, energy & fuels, forensics, materials science, biopharmaceutical, proteomics, metabolomics).
  • Plastic springs provide shock absorption, preventing syringe damage.
  • Polypropylene construction prevents compound adhesion to surfaces.