Learn More
Microsolv Technology Corporation Insert Rsa Deactivated
Catalog No. 5001452060
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, Surface Treated, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. RSA-Pro Brand. 100/PK.
Catalog No. 50-014-52060 Supplier Microsolv Technology Corporation Supplier No. 9502S02NDRS
Inserts, Low Volume for Vials, Glass. Clear, Surface Treated, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. RSA-Pro Brand. 100/PK.