Learn More
Microsolv Technology Corporation Insert Rsa Glass Spring
Catalog No. 5001452061
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic springs attached. For use in 2ml autosampler vials. RSA Brand. 100/PK.
Catalog No. 50-014-52061 Supplier Microsolv Technology Corporation Supplier No. 9502S02NRS
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic springs attached. For use in 2ml autosampler vials. RSA Brand. 100/PK.