Learn More
Hach Company Insertion Mount Analog SST
Supplier:  Hach Company 5646450
Encompass
Insertion Mounting-Analog-1 1/2in SSTIncludes a 1 5in ball valve in 316 stainless steel 1 5in NPT close nipple sensor adapter with two FKM/FPM For use with Hach analog pHD sensors
Catalog No. 50-002-07085