Learn More
Hach Company Insertion Mount Digital CPVC
Supplier:  Hach Company 5646300
Encompass
Insertion Mounting-Digital-1 1/2in CPVCIncludes a 1 5in ball valve in CPVC 1 5in NPT close nipple sensor adapter with two FKM/FPM For use with Hach digital pHD sensors
Catalog No. 50-002-07267