Learn More
Hach Company Solitax Inline sc Turbidity and Suspended Solids Insertion Probe with Wiper, Stainless Steel (LXV424.99.00100)
Catalog No. NC3315478 Shop All Hach Company Products
Encompass
The Solitax Inline sc insertion probe is a stainless steel, in-pipe sensor for continuous, color-independent measurement of turbidity and suspended solids using an infrared duo scattered light technique. It includes an automatic wiper for reduced maintenance and is suited for drinking water, wastewater, and industrial process monitoring. The probe supports digital controller integration and remote data management.
  • High-precision, color-independent turbidity and suspended solids measurement
  • Automatic wiper reduces fouling and maintenance
  • Stainless steel construction for corrosion resistance
  • Wide measurement range: turbidity 0.001-4000 NTU; TSS 0.001 mg/L-50 g/L
  • IP68 rating for reliable operation in wet environments
  • Insertion mounting for in-pipe use
  • Adjustable response time (T90 1-300 s)
  • Standard 10 m cable; sensor-to-controller up to 100 m
  • Certified to DIN EN ISO 7027 and MCERTS
Catalog No. NC3315478 Supplier Hach Company Supplier No. LXV424.99.00100
Add to Cart
Add to Cart
Request a Quote
The Solitax Inline sc insertion probe is a stainless steel, in-pipe sensor for continuous, color-independent measurement of turbidity and suspended solids using an infrared duo scattered light technique. It includes an automatic wiper for reduced maintenance and is suited for drinking water, wastewater, and industrial process monitoring. The probe supports digital controller integration and remote data management.
  • High-precision, color-independent turbidity and suspended solids measurement
  • Automatic wiper reduces fouling and maintenance
  • Stainless steel construction for corrosion resistance
  • Wide measurement range: turbidity 0.001-4000 NTU; TSS 0.001 mg/L-50 g/L
  • IP68 rating for reliable operation in wet environments
  • Insertion mounting for in-pipe use
  • Adjustable response time (T90 1-300 s)
  • Standard 10 m cable; sensor-to-controller up to 100 m
  • Certified to DIN EN ISO 7027 and MCERTS