Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Inserts for 1.5 mL Large Opening Vials

Catalog No. 111009060 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Insert Type:
Conical
Conical with Bottom Spring
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Insert Type
11-100-9060 Conical
11-101-0584 Conical with Bottom Spring
11-100-6743 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-9060 Supplier MilliporeSigma™ Supelco™ Supplier No. 854988
Add to Cart
Add to Cart

Allow the use of smaller sample volumes with standard autosampler vials.

  • Vial inserts for 1.5 mL large opening vials [0.1 mL, clear glass conical (with bottom spring)] allow the use of smaller sample volumes with standard autosampler vials.
  • Inserts are generally chosen based on the inner diameter of the vial neck.
  • Large opening vials (6 mm ID) require a 6 mm OD insert.
  • Clear glass conical inserts used with CTC LC PAL autosamplers.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Glass
Volume (Metric) 0.1 mL
Insert Type Conical
Quantity 1000 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.