Learn More
Microsolv Technology Corporation Inserts 350Ul 6X31Mm Flat Bot
Catalog No. 5001452072
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 350ul fill volume with a flat bottom and 6x31mm outer dimensions. For use in 2ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52072 Supplier Microsolv Technology Corporation Supplier No. 9502S05N
Inserts, Low Volume for Vials, Glass. Clear, 350ul fill volume with a flat bottom and 6x31mm outer dimensions. For use in 2ml autosampler vials. AQ Brand. 100/PK.