Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Inserts for Cryo.s Storage Vials

Catalog No. 07000984 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-984 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-984 Supplier Greiner Bio-One Supplier No. 304171P

missing translation for 'validMainframeMsg'

Used for labeling and identification purposes.

  • Cap for labeling and easy identification

Service Plan

No

TRUSTED_SUSTAINABILITY

Specifications

Color White
Sterility Non-sterile
For Use With (Application) Cap inserts for labeling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.