Learn More
Microsolv Technology Corporation Inserts Glass 100Ul 5X29Mm
Catalog No. 5001452398
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 200ul fill volume with a conical point and 5x29mm outer dimensions with plastic springs attached. For use in 8mm, 2ml autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52398 Supplier Microsolv Technology Corporation Supplier No. 9582S02N
Inserts, Low Volume for Vials, Glass. Clear, 200ul fill volume with a conical point and 5x29mm outer dimensions with plastic springs attached. For use in 8mm, 2ml autosampler vials. MicroSolv Brand. 100/PK.