Learn More
Microsolv Technology Corporation Inserts Glass 200Ul 6X29 Sprgs
Catalog No. 5001451808
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 200ul fill volume with a conical, high precision point and 6x29mm outer dimensions with plastic spring attached. For use in 1ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-51808 Supplier Microsolv Technology Corporation Supplier No. 9500104N
Inserts, Low Volume for Vials, Glass. Clear, 200ul fill volume with a conical, high precision point and 6x29mm outer dimensions with plastic spring attached. For use in 1ml autosampler vials. AQ Brand. 100/PK.