Learn More
Microsolv Technology Corporation Inserts, in Racks with a Base. Includes 96, Clear, 500ul Glass Low Volume Precision Inserts with wide conical points. For use in the Micro-Sample Management System. 1 EA.
Catalog No. 502270536
Encompass_Preferred
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System, Pre-Loaded in a Rack with a Base. These Inserts have a wide conical point for fast sample filling and minimizes bubble formation. Sealing Film or Mats sold separately.
This item is not returnable. View return policy
Catalog No. 50-227-0536 Supplier Microsolv Technology Corporation Supplier No. 95296U2D500
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System, Pre-Loaded in a Rack with a Base. These Inserts have a wide conical point for fast sample filling and minimizes bubble formation. Sealing Film or Mats sold separately.