Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 100ul fill volume with a conical, precision point and 5x30mm outer dimensions with plastic spring attached. For use in 8mm, Push Top, 1ml autosampler vials. 100/PK.
Catalog No. 502270523
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features thicker glass walls than other similar inserts and a precision point conical tip for maximum recovery of sample. Residual volume is 2ul with most autosamplers.Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use. This product is manufactured in an ISO 9001:2015 Certified facility. Can be used with 8x40mm, "96 position" vials.
This item is not returnable. View return policy
Catalog No. 50-227-0523 Supplier Microsolv Technology Corporation Supplier No. 9500103N
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features thicker glass walls than other similar inserts and a precision point conical tip for maximum recovery of sample. Residual volume is 2ul with most autosamplers.Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use. This product is manufactured in an ISO 9001:2015 Certified facility. Can be used with 8x40mm, "96 position" vials.