Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 200ul fill volume with a conical, precision point and 6x29mm outer dimensions. For use in 8mm, Push Top, 1ml autosampler vials. 100/PK.
Catalog No. 502270520
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a precision point conical tip for maximum recovery of sample. Residual volume is 2ul with most autosamplers. This product is manufactured in an ISO 9001:2015 Certified facility. Spring may be required, sold separately, Can be used with 8x40mm, "96 position" vials.
This item is not returnable. View return policy
Catalog No. 50-227-0520 Supplier Microsolv Technology Corporation Supplier No. 9500101N
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a precision point conical tip for maximum recovery of sample. Residual volume is 2ul with most autosamplers. This product is manufactured in an ISO 9001:2015 Certified facility. Spring may be required, sold separately, Can be used with 8x40mm, "96 position" vials.