Learn More
Microsolv Technology Corporation Inserts Prec Deac Spg
Catalog No. 5001452059
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52059 Supplier Microsolv Technology Corporation Supplier No. 9502S02ND
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic spring attached. For use in 2ml autosampler vials. AQ Brand. 100/PK.