Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume 
Catalog No. 5001452058
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic springs attached. For use in 2ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52058 Supplier Microsolv Technology Corporation Supplier No. 9502S02N
Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a conical, precision point and 6x29mm outer dimensions with plastic springs attached. For use in 2ml autosampler vials. AQ Brand. 100/PK.