Learn More
Microsolv Technology Corporation Inserts Silan Glass 100Ul 5X30
Catalog No. 5001451807
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 100ul fill volume with a conical, precision point and 5x30mm outer dimensions with plastic spring attached. For use in 8mm, Push Top, 1ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-51807 Supplier Microsolv Technology Corporation Supplier No. 9500103ND
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 100ul fill volume with a conical, precision point and 5x30mm outer dimensions with plastic spring attached. For use in 8mm, Push Top, 1ml autosampler vials. AQ Brand. 100/PK.