Learn More
Microsolv Technology Corporation Inserts Silan Glass 200Ul 6X29
Catalog No. 5001451803
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 200ul fill volume with a conical, precision point and 6x29mm outer dimensions. For use in 8mm, Push Top, 1ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-51803 Supplier Microsolv Technology Corporation Supplier No. 9500101ND
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 200ul fill volume with a conical, precision point and 6x29mm outer dimensions. For use in 8mm, Push Top, 1ml autosampler vials. AQ Brand. 100/PK.