Learn More
Microsolv Technology Corporation Inserts Silan Glass 200Ul 6X29
Supplier:  Microsolv Technology Corporation 9500104ND
Encompass_Preferred
Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 200ul fill volume with a conical, high precision point and 6x29mm outer dimensions with plastic spring attached. For use in 8mm, Push Top,1ml autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-51809