Learn More
Beckman Coulter 373698 Biomek Rack Insert F 12mm
Catalog No. NC9604841
Encompass_Preferred
The Beckman Coulter 373698 Biomek Rack Insert F 12mm is a rack insert designed for use with Beckman Coulter Biomek liquid handling systems. It is made of polypropylene and has 12mm diameter openings to accommodate various labware. The insert allows for organized storage and access to tubes and vials during automated protocols on the Biomek platforms. With dimensions of 127 x 86 mm, this rack insert provides a standard footprint that is compatible with Biomek 1000, 2000, 3000, 4000 series liquid handlers. The rigid construction securely holds labware in place for reliable operation.
Catalog No. NC9604841 Supplier Beckman Coulter Supplier No. 373698
The Beckman Coulter 373698 Biomek Rack Insert F 12mm is a rack insert designed for use with Beckman Coulter Biomek liquid handling systems. It is made of polypropylene and has 12mm diameter openings to accommodate various labware. The insert allows for organized storage and access to tubes and vials during automated protocols on the Biomek platforms. With dimensions of 127 x 86 mm, this rack insert provides a standard footprint that is compatible with Biomek 1000, 2000, 3000, 4000 series liquid handlers. The rigid construction securely holds labware in place for reliable operation.