Learn More
Beckman Coulter Inserts, Tube 13mm Biomek
Catalog No. NC9038629
Encompass_Preferred
The Beckman Coulter 13mm Inserts Tube is a plastic tube insert designed for use with Beckman Coulter Biomek automated liquid handling systems. It has an outer diameter of 13mm and an inner diameter of 10mm. The tube is made from polypropylene and has a working volume of 3mL. It can withstand temperatures from -20°C to +120°C. The tube has a conical bottom and comes sterile and individually wrapped. It is compatible with Beckman Coulter Biomek 1000, 2000, 3000, 3000 Laboratory Automation Workstations.
Catalog No. NC9038629 Supplier Beckman Coulter Supplier No. 373697
The Beckman Coulter 13mm Inserts Tube is a plastic tube insert designed for use with Beckman Coulter Biomek automated liquid handling systems. It has an outer diameter of 13mm and an inner diameter of 10mm. The tube is made from polypropylene and has a working volume of 3mL. It can withstand temperatures from -20°C to +120°C. The tube has a conical bottom and comes sterile and individually wrapped. It is compatible with Beckman Coulter Biomek 1000, 2000, 3000, 3000 Laboratory Automation Workstations.