Learn More
Aviva Systems Biology Corp INSIG1 Antibody - middle region (ARP40162_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40162_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INSIG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human INSIG1. Alternate names: CL-6, MGC1405, CL6ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57311