Learn More
Aviva Systems Biology Corp INSIG2 Antibody - N-terminal region (ARP40165_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40165_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INSIG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human INSIG2. Alternate names: MGC26273ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53177