Learn More
Marketlab Insight GynoCart
Description
- Converts any ED stretcher into an exam table
- Integrated stirrups for proper and dignified patient positioning
- Fold-out pad for patient comfort and cart stability
- Height-adjustable exam stool with casters is stored inside cart
- Extendable storage shelves provide easy access to supplies
- Moves effortlessly on four 5 in. casters; two lockable
- Doors with magnetic closures can accommodate tamper-resistant locks
- More lower-cost than a full gynecological bed
- Model with light has 15 ft. power cord with metal cord wrap to organize while not in use
Specifications
Specifications
| Dimensions (D x W x H) | 25.25 x 38 x 35.62 in. |
| Material | Aluminum |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.