Learn More
Thermo Scientific™ Insight™ Pro Software
Catalog No. 14385511 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Insight Pro Security Software
Insight Pro Software
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-385-511 Insight Pro Security Software
14-385-512 Insight Pro Software
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-385-511 Supplier Thermo Scientific™ Supplier No. 837004000
Add to Cart
Add to Cart
Request a Quote
Streamline your routine, complex and advanced routines and workflows with Thermo Scientific™ Insight™ Pro Software. Prevent costly sampling and measurement errors with prompting, step-by-step workflows.
Streamline your routine, complex and advanced routines and workflows with Thermo Scientific™ Insight™ Pro Software. Prevent costly sampling and measurement errors with prompting, step-by-step workflows. Add Thermo Scientific™ Insight™ Pro Security Software to achieve compliance with current FDA 21 CFR Part 11 regulations.
When installation and operational qualifications (IQ/OQ) are required, Thermo Scientific™ Validator™ for Insight Pro Software provides all the documentation and reference materials needed to complete IQ/OQ for Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers.
Make your lab time more productive with Insight Pro Software
  • Reduce training time - Easy to teach and learn, optimized user interface with prompting software defines clear step-by- step workflows
  • Prevent costly sampling and measurement errors - Create dedicated workflows for complex assay and transform multistep workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current - Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
Streamlined QA/QC Analysis:
  • Automated result calculation options eliminate manual calculations and reduce user error.
  • Detailed graphical displays of calibration curves and sample run charts provide quick visual checks of system suitability and data trends.
  • Control limits and automated pass/fail interpretation provide immediate compliance results.
  • Scanning Quant includes ability to perform multiple calculations and extract parameters to create a complete analytical picture, improving method development experience.
Simplified Research Applications:
  • Multitude of data collection options and graphing features.
  • Pre-programmed peak analysis and convenient spectral processing tools aid in sample identification and quantitation for scanning measurements.
  • Multi-staged curve fitting and consecutive reaction modes provide comprehensive data fitting options for kinetics experiments.
  • Sophisticated math analysis functions, including derivatives and smoothing allow custom data display.
  • Merge workbooks feature allows quick and convenient comparison of data from multiple experiments.
Integrated Accessories:
  • Automated accessories allow complete control for superior performance with advanced sample handling requirements.
  • Calibration and alignment tools ensure accessories are performing optimally.
  • Supported temperature control accessories control and monitor reaction temperature in eight positions.
  • Programmable sipper, cell changer or autosampler accessory performs high-throughput activities with ease.
  • Hands-free performance verification and accessory calibration modules are designed for the SMART Sipper or Linear 8-Cell Changer accessory.
Comply with US FDA 21 CFR Part 11:
To ensure the security and integrity of your data and to help your lab comply with 21 CFR Part 11 or other regulations, add Insight Pro Security Software with which pairs Insight Pro Software with Thermo Scientific™; Security Suite Software.
  • Control user account access
  • Set and manage security policies to ensure proper data handling.
  • Customize and enforce digital signatures to establish responsibility.
  • Log and view security events
  • Electronic audit trails for a reliable electronic documentation.
  • Database storage ensures to maintain electronic records for FDA submissions
Instrument Qualification:
Validator for Insight Pro Software includes:
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance
  • Installation and instrument certification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Supports current 21 CFR Part 11 compliance
Type Insight Pro Security Software
For Use With (Equipment) For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers