Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Insight Storage Cart, 5 in. Casters

Catalog No. 22505058
Change view
Click to view available options
Locking Mechanism:
Electronic
Key
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Locking Mechanism
22-505-058 Key
22-505-094 Electronic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-505-058 Supplier Marketlab Supplier No. 43350
Add to Cart
Add to Cart

Perfect Balance of Storage Capacity and Workspace

  • Five full-extension drawers maximize storage capacity
  • Stain-resistant worksurface
  • Additional pull-out worksurface expands workspace without increasing cart's footprint
  • Aluminum frame doubles as rail system for adding accessories
  • Handles on each side of cart allow you to move cart from any angle
  • Lightweight design and durable medical-grade casters for easy and quiet movement over any surface
  • 1-year limited warranty

Specifications

Product Type Phlebotomy Cart
Dimensions (D x W x H) 19 x 25.25 x 36.75 in.
Locking Mechanism Key
Material Aluminum
No. of Shelves None
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.