Learn More
Marketlab Insight Storage Cart, 5 in. Casters
Catalog No. 22505094
Change view
Click to view available options
Locking Mechanism:
Electronic
Key
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Locking Mechanism
22-505-094 Electronic
22-505-058 Key
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-505-094 Supplier Marketlab Supplier No. 43350E
Add to Cart
Add to Cart
Perfect Balance of Storage Capacity and Workspace
  • Five full-extension drawers maximize storage capacity
  • Stain-resistant worksurface
  • Additional pull-out worksurface expands workspace without increasing cart's footprint
  • Aluminum frame doubles as rail system for adding accessories
  • Handles on each side of cart allow you to move cart from any angle
  • Lightweight design and durable medical-grade casters for easy and quiet movement over any surface
  • 1-year limited warranty
Specifications
Product Type Phlebotomy Cart
Dimensions (D x W x H) 19 x 25.25 x 36.75 in.
Locking Mechanism Electronic
Material Aluminum
No. of Shelves None