Learn More
MSC Insize Dial Caliper, 0-4 in, 0.001 in Graduation, Stainless Steel
Catalog No. 5000537584
Encompass
This dial caliper features a 4-inch measurement range and a dial graduation of 0.001 inches. It offers a range of 0.100 inches per revolution and an accuracy of +/-0.0015 inches. The caliper is calibrated and constructed with stainless steel jaws and a stainless steel body. It comes with a manufacturer inspection certificate traceable to NIST and a hard plastic case. The dial face is white, and the dial case is gray. The caliper is shockproof.
  • Shockproof design provides durability and protection.
  • Constructed from stainless steel for robust construction and longevity.
  • Supplied with manufacturer inspection certificate traceable to NIST, guaranteeing accuracy and compliance.
  • Includes a hard plastic case for protection and convenient storage.
Catalog No. 50-005-37584 Supplier MSC Supplier No. 28731057
This dial caliper features a 4-inch measurement range and a dial graduation of 0.001 inches. It offers a range of 0.100 inches per revolution and an accuracy of +/-0.0015 inches. The caliper is calibrated and constructed with stainless steel jaws and a stainless steel body. It comes with a manufacturer inspection certificate traceable to NIST and a hard plastic case. The dial face is white, and the dial case is gray. The caliper is shockproof.
  • Shockproof design provides durability and protection.
  • Constructed from stainless steel for robust construction and longevity.
  • Supplied with manufacturer inspection certificate traceable to NIST, guaranteeing accuracy and compliance.
  • Includes a hard plastic case for protection and convenient storage.