Learn More
MSC Insize Radius Gage Set, Medium Carbon Steel, 17/32-1 in
Catalog No. 5000533954
Encompass
This radius gage set is designed for measuring radii from 17/32 to 1 " Made from medium carbon steel, it includes 16 pieces, offering a comprehensive range for various applications. It provides an accuracy of ±.0017 inch, suitable for general use.
  • Made of medium carbon steel
  • Measures radii from 17/32-1 in
  • Includes 16 pieces
  • Individual leaf radii: 17/32 in, 9/16 in, 19/32 in, 5/8 in, 21/32 in, 11/16 in, 23/32 in, 3/4 in, 25/32 in, 13/16 in, 27/32 in, 7/8 in, 29/32 in, 15/16 in, 31/32 in, 1 in
  • Accuracy of ±.0017 in
  • Features low accuracy for general measurement tasks
Catalog No. 50-005-33954 Supplier MSC Supplier No. 28795201
This radius gage set is designed for measuring radii from 17/32 to 1 " Made from medium carbon steel, it includes 16 pieces, offering a comprehensive range for various applications. It provides an accuracy of ±.0017 inch, suitable for general use.
  • Made of medium carbon steel
  • Measures radii from 17/32-1 in
  • Includes 16 pieces
  • Individual leaf radii: 17/32 in, 9/16 in, 19/32 in, 5/8 in, 21/32 in, 11/16 in, 23/32 in, 3/4 in, 25/32 in, 13/16 in, 27/32 in, 7/8 in, 29/32 in, 15/16 in, 31/32 in, 1 in
  • Accuracy of ±.0017 in
  • Features low accuracy for general measurement tasks